ENA428072539 Enamine

(5S)-2,2,5-trimethylmorpholine

  • MDL number MFCD28023686
  • Cas Number : 1807921-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178113

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178113
Availability details for
Quantity
  • Availability
  • Add to Cart