ENA428072541 Enamine

1-(1-methyl-1H-imidazol-5-yl)ethan-1-one hydrochloride

  • MDL number MFCD28023656
  • Cas Number : 20970-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178125

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178125
Availability details for
Quantity
  • Availability
  • Add to Cart