ENA428072542 Enamine

1-(1-methyl-1H-imidazol-2-yl)piperidin-4-amine dihydrochloride

  • MDL number MFCD28023657
  • Cas Number : 1790137-35-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178128
Availability details for
Quantity
  • Availability
  • Add to Cart