ENA428072546 Enamine

2-[(tert-butyldimethylsilyl)oxy]propane-1-sulfonyl chloride

  • MDL number MFCD28064274
  • Cas Number : 1643683-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-178166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178166
Availability details for
Quantity
  • Availability
  • Add to Cart