ENA428072556 Enamine

3-cyclopropyl-5-[(2R,4S)-4-methoxypyrrolidin-2-yl]-1,2,4-oxadiazole hydrochloride

  • MDL number MFCD28023664
  • Cas Number : 1807939-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-178290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178290
Availability details for
Quantity
  • Availability
  • Add to Cart