ENA428072558 Enamine

5-(aminomethyl)-2-fluoro-N-(propan-2-yl)benzene-1-sulfonamide hydrochloride

  • MDL number MFCD28023667
  • Cas Number : 1803605-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-178296

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-178296
Availability details for
Quantity
  • Availability
  • Add to Cart