ENA428072563 Enamine

1-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]-1,4-diazepane

  • MDL number MFCD28023727
  • Cas Number : 946409-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160513

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160513
Availability details for
Quantity
  • Availability
  • Add to Cart