ENA428072577 Enamine

1-chloro-2-(chloromethyl)-3-methylbenzene

  • MDL number MFCD12025220
  • Cas Number : 55676-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-170811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-170811
Availability details for
Quantity
  • Availability
  • Add to Cart