ENA428072631 Enamine

(4-bromo-3,5-dichlorophenyl)methanol

  • MDL number MFCD28023735
  • Cas Number : 1788545-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-173729

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-173729
Availability details for
Quantity
  • Availability
  • Add to Cart