ENA428072651 Enamine

tert-butyl 4-sulfamoylpiperidine-1-carboxylate

  • MDL number MFCD28023737
  • Cas Number : 1415929-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-174058

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-174058
Availability details for
Quantity
  • Availability
  • Add to Cart