ENA428072658 Enamine

ethyl 2-chloro-2-[2-(2,4-difluorophenyl)hydrazin-1-ylidene]acetate

  • Cas Number : 1264087-24-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-174317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-174317
Availability details for
Quantity
  • Availability
  • Add to Cart