ENA428072663 Enamine

ethyl 3-methyl-2-methylidenebutanoate

  • MDL number MFCD28023687
  • Cas Number : 68834-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-130952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-130952
Availability details for
Quantity
  • Availability
  • Add to Cart