ENA428072737 Enamine

{[4-(bromomethyl)phenyl]methoxy}(tert-butyl)dimethylsilane

  • MDL number MFCD28023764
  • Cas Number : 125320-20-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-175534

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-175534
Availability details for
Quantity
  • Availability
  • Add to Cart