ENA428072759 Enamine

2-(2,2,2-trifluoroethoxy)-6-(trifluoromethyl)pyridine-3-carbonitrile

  • MDL number MFCD23356219
  • Cas Number : 1499589-54-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-175797
Availability details for
Quantity
  • Availability
  • Add to Cart