ENA428072784 Enamine

[4-(cyclopropylmethoxy)phenyl]methanol

  • MDL number MFCD11190318
  • Cas Number : 712313-61-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176107
Availability details for
Quantity
  • Availability
  • Add to Cart