ENA428072803 Enamine

1',2'-dihydrospiro[cyclobutane-1,3'-indole] hydrochloride

  • MDL number MFCD28023681
  • Cas Number : 1689557-88-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176323

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176323
Availability details for
Quantity
  • Availability
  • Add to Cart