ENA428072825 Enamine

4-[4-(trifluoromethyl)phenyl]-1H-imidazol-2-amine

  • MDL number MFCD21717737
  • Cas Number : 37148-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176454

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176454
Availability details for
Quantity
  • Availability
  • Add to Cart