ENA428072832 Enamine

3-(pyrimidin-4-yl)propanoic acid

  • MDL number MFCD06796207
  • Cas Number : 819850-17-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176496
Availability details for
Quantity
  • Availability
  • Add to Cart