ENA428072835 Enamine

4-(2-bromophenyl)-3-methyl-1H-pyrazole

  • MDL number MFCD22034891
  • Cas Number : 1368048-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176535
Availability details for
Quantity
  • Availability
  • Add to Cart