ENA428072840 Enamine

4-cyclobutylbenzoic acid

  • MDL number MFCD19440000
  • Cas Number : 3158-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176558
Availability details for
Quantity
  • Availability
  • Add to Cart