ENA428072843 Enamine

1-ethyl-3-propyl-2,3-dihydro-1H-1,3-benzodiazol-2-imine

  • MDL number MFCD02993919
  • Cas Number : 487017-58-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176562
Availability details for
Quantity
  • Availability
  • Add to Cart