ENA428072845 Enamine

spiro[2.3]hexan-5-one

  • MDL number MFCD23884029
  • Cas Number : 20061-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-176564

Safety Information

signalword Warning
pcodes P210,P233,P240,P241,P242,P243,P261,P264,P271,P280,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
hcodes Xi
MSDS https://www.enaminestore.com/catalog/EN300-176564
pictograms GHS02,GHS07
hstatements H226,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart