ENA428072852 Enamine

4-cyclopropyl-6-methylpyrimidin-2-amine

  • MDL number MFCD16769143
  • Cas Number : 92238-61-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176584

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176584
Availability details for
Quantity
  • Availability
  • Add to Cart