ENA428072890 Enamine

2-[(1,1,1-trifluoropropan-2-yl)oxy]pyridine-3-carboxylic acid

  • MDL number MFCD21737558
  • Cas Number : 1502732-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176705

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176705
Availability details for
Quantity
  • Availability
  • Add to Cart