ENA428072891 Enamine

9H-fluorene-9-carbonitrile

  • MDL number MFCD00604077
  • Cas Number : 1529-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-152
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176706

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176706
Availability details for
Quantity
  • Availability
  • Add to Cart