ENA428072898 Enamine

methyl 2-[3-(aminomethyl)phenyl]-1,3-thiazole-4-carboxylate hydrobromide

  • MDL number MFCD28023602
  • Cas Number : 1798732-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176755
Availability details for
Quantity
  • Availability
  • Add to Cart