ENA428072902 Enamine

1-ethyl-6-methyl-1H-1,3-benzodiazole

  • MDL number MFCD18825717
  • Cas Number : 26530-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176772
Availability details for
Quantity
  • Availability
  • Add to Cart