ENA428072921 Enamine

1-(3-bromophenyl)-4,4,4-trifluorobutan-2-amine

  • MDL number MFCD14657049
  • Cas Number : 1250792-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-176806

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176806
Availability details for
Quantity
  • Availability
  • Add to Cart