ENA428072922 Enamine

5-methyl-1-{[4-(trifluoromethyl)phenyl]methyl}-1H-1,2,3-triazol-4-amine

  • MDL number MFCD22703226
  • Cas Number : 1528795-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176809
Availability details for
Quantity
  • Availability
  • Add to Cart