ENA428072928 Enamine

methyl 1-ethyl-1H-pyrazole-3-carboxylate

  • MDL number MFCD09701705
  • Cas Number : 89943-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176849
Availability details for
Quantity
  • Availability
  • Add to Cart