ENA428072936 Enamine

3,4-dihydro-2H-1-benzothiopyran-4-thiol

  • MDL number MFCD10692119
  • Cas Number : 127661-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-176909
Availability details for
Quantity
  • Availability
  • Add to Cart