ENA428072940 Enamine

4-methyl-3,4-dihydro-2H-1,4-benzoxazine-6-carboxylic acid

  • MDL number MFCD12025919
  • Cas Number : 532391-92-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-176927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176927
Availability details for
Quantity
  • Availability
  • Add to Cart