ENA428072951 Enamine

2-(oxolan-2-yl)acetyl chloride

  • MDL number MFCD21128707

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-176969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176969
Availability details for
Quantity
  • Availability
  • Add to Cart