ENA428072954 Enamine

4,7-dimethyl-1,3-dihydro-2-benzofuran-1,3-dione

  • MDL number MFCD00094951
  • Cas Number : 5463-50-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-145
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-176972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-176972
Availability details for
Quantity
  • Availability
  • Add to Cart