ENA428072964 Enamine

3,4-dichloro-5-iodobenzoic acid

  • MDL number MFCD24107200
  • Cas Number : 1507268-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177006

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177006
Availability details for
Quantity
  • Availability
  • Add to Cart