ENA428072972 Enamine

N-(6-aminohexyl)guanidine; sulfuric acid

  • MDL number MFCD01632444
  • Cas Number : 1401846-54-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data H2SO4
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177022
Availability details for
Quantity
  • Availability
  • Add to Cart