ENA428072974 Enamine

6-[ethyl(methyl)amino]pyridine-3-carbaldehyde

  • MDL number MFCD12403453
  • Cas Number : 1216170-74-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177030

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177030
Availability details for
Quantity
  • Availability
  • Add to Cart