ENA428072977 Enamine

[4-(trimethylsilyl)phenyl]methanethiol

  • MDL number MFCD06200763
  • Cas Number : 57337-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177046
Availability details for
Quantity
  • Availability
  • Add to Cart