ENA428072985 Enamine

{3-[(1-methoxypropan-2-yl)oxy]phenyl}boronic acid

  • MDL number MFCD19654447
  • Cas Number : 1350317-80-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177063

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177063
Availability details for
Quantity
  • Availability
  • Add to Cart