ENA428072986 Enamine

N-(2-bromo-6-fluorophenyl)-4,5-dihydro-1H-imidazol-2-amine

  • MDL number MFCD00868606
  • Cas Number : 65896-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177066
Availability details for
Quantity
  • Availability
  • Add to Cart