ENA428072990 Enamine

N2-[(3,5-dimethyl-1,2-oxazol-4-yl)methyl]-1H-1,3-benzodiazole-2,5-diamine

  • MDL number MFCD22920797
  • Cas Number : 1499805-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177073
Availability details for
Quantity
  • Availability
  • Add to Cart