ENA428072991 Enamine

6-methylpyridine-3-sulfonamide

  • MDL number MFCD18819130
  • Cas Number : 34082-13-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 151-155
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177075

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177075
Availability details for
Quantity
  • Availability
  • Add to Cart