ENA428073000 Enamine

1,3-dichloro-2-ethynylbenzene

  • MDL number MFCD16616267
  • Cas Number : 6575-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177115
Availability details for
Quantity
  • Availability
  • Add to Cart