ENA428073011 Enamine

1-methoxycyclohexane-1-carbaldehyde

  • MDL number MFCD24684644
  • Cas Number : 761411-22-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. -20

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177129
Availability details for
Quantity
  • Availability
  • Add to Cart