ENA428073015 Enamine

1-(3-chlorophenyl)-5-ethyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD28064252
  • Cas Number : 1795184-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177157
Availability details for
Quantity
  • Availability
  • Add to Cart