ENA428073018 Enamine

1-{1-oxaspiro[4.5]decan-2-yl}methanamine hydrochloride

  • MDL number MFCD28023684
  • Cas Number : 1795330-56-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177165
Availability details for
Quantity
  • Availability
  • Add to Cart