ENA428073020 Enamine

5,5-dimethyl-1,3-diazinane-2,4-dione

  • MDL number MFCD19217287
  • Cas Number : 26239-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260-261
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177201

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177201
Availability details for
Quantity
  • Availability
  • Add to Cart