ENA428073021 Enamine

6-[methyl(propan-2-yl)amino]pyridine-3-carbaldehyde

  • MDL number MFCD23952419
  • Cas Number : 1526498-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-177202

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177202
Availability details for
Quantity
  • Availability
  • Add to Cart