ENA428073022 Enamine

3-[(3-aminophenyl)methyl]-3,4-dihydroquinazolin-4-one

  • MDL number MFCD12631583
  • Cas Number : 1042796-67-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-177203
Availability details for
Quantity
  • Availability
  • Add to Cart