ENA428073028 Enamine

5-methyl-4-nitrofuran-2-carboxylic acid

  • MDL number MFCD15144147
  • Cas Number : 326867-81-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-177237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-177237
Availability details for
Quantity
  • Availability
  • Add to Cart